Skip to Content
Merck
MacConkey Broth ready-to-use, bottle volume 100 mL , filling volume

MacCONKEY broth

MacConkey Broth ready-to-use, bottle volume 100 mL , filling volume

Products

Product Category
Brand
Color
pH Value
Physical Form
Agency
Chemical Composition
Capacity (mL)
Volume (ml)
Quality Segment
Closure Type
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique