
Lysophosphatidylcholine (Soy)

Synonym(s):
L-α-Lysophosphatidylcholine from Glycine max (soybean), 1-Acyl-sn-glycero-3-phosphocholine, L-α-Lysolecithin
CAS Number:
Products
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing