
Loratadine Related Compound B

Synonym(s):
8-Chloro-6,11-dihydro-11-(1-methyl-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, N-Methyldesloratadine, 8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
Empirical Formula (Hill Notation):
C20H21ClN2
CAS Number:
Molecular Weight:
324.85
Products
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing