Skip to Content
Merck
Loganin analytical standard

Loganin

Loganin analytical standard
Synonym(s):
7-Hydroxy 6-desoxyverbenalin, Loganoside
Empirical Formula (Hill Notation):
C17H26O10
CAS Number:
Molecular Weight:
390.38
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing