
L-Lysine-1-13C hydrochloride

Linear Formula:
H2N(CH2)4CH(NH2)13CO2H·HCl
CAS Number:
Molecular Weight:
183.64
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing