Skip to Content
Merck
Lithium cobalt(III) oxide 99.8% trace metals basis

Lithium cobalt(III) oxide

Lithium cobalt(III) oxide 99.8% trace metals basis
Synonym(s):
Lithium cobaltite
Linear Formula:
LiCoO2
CAS Number:
Molecular Weight:
97.87
EC Number:

Products

Shipping
Product Category
Sustainability
Feature
Formula Weight
Physical Form
Purity
Chemical Composition
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing