
L-Glutamic acid di-tert-butyl ester hydrochloride

Empirical Formula (Hill Notation):
C13H25NO4 · HCl
CAS Number:
Molecular Weight:
295.80
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Amino Acid
Showing 1-1 of 1