
L-α-Lysophosphatidylcholine

Synonym(s):
3-sn-Lysophosphatidylcholine, L-α-Lysolecithin
Products
Product Category
Brand
Biological Source
Physical Form
Purity
Tissue
Chemical Composition
Quality Segment
Functional Group
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing