Skip to Content
Merck
L-Alanine-15N 98 atom % 15N, endotoxin tested

L-Alanine-15N

L-Alanine-15N 98 atom % 15N, endotoxin tested
Synonym(s):
(S)-2-Aminopropionic acid-15N, L-α-Aminopropionic acid-15N, 15N Labeled L-alanine
Linear Formula:
CH3CH(15NH2)CO2H
CAS Number:
Molecular Weight:
90.09

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing