Skip to Content
Merck
L-165,041 ≥98% (HPLC), powder

L-165,041

L-165,041 ≥98% (HPLC), powder
Synonym(s):
L-165,041, 4-[3-(4-Acetyl-3-hydroxy-2-propylphenoxy)propoxy]phenoxy­acetic acid
Empirical Formula (Hill Notation):
C22H26O7
CAS Number:
Molecular Weight:
402.44

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment