Isopropyl alcohol
Synonym(s):
2-Propanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol
Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Products
Shipping
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3