
Iobenguane sulfate

Synonym(s):
m-Iodobenzylguanidine hemisulfate salt, MIBG
Linear Formula:
C8H10IN3 · 1/2H2SO4
CAS Number:
Molecular Weight:
324.13
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing