Skip to Content
Merck
vibrant-m

Interference Mix 7 (For Qualitative Use Only)

vibrant-m
Synonym(s):
Qualitative Interference Mixture

Products

Brand
Manufacturer
Physical Form
Format
Chemical Composition
Application
Package Size
Special Grade
Technique
Showing 1-1 of 1