![IAEA-S-2 (sulfur isotopes in silver sulfide) NIST® RM 8555](/deepweb/assets/sigmaaldrich/product/structures/229/789/2728cbce-78e4-444c-8f38-3cff2285168d/640/2728cbce-78e4-444c-8f38-3cff2285168d.png)
IAEA-S-2 (sulfur isotopes in silver sulfide)
![IAEA-S-2 (sulfur isotopes in silver sulfide) NIST® RM 8555](/deepweb/assets/sigmaaldrich/product/structures/229/789/2728cbce-78e4-444c-8f38-3cff2285168d/640/2728cbce-78e4-444c-8f38-3cff2285168d.png)
Synonym(s):
Silver sulfide
Empirical Formula (Hill Notation):
Ag2S
CAS Number:
Molecular Weight:
247.80
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Isotope
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing