Skip to Content
Merck
Hydrocortisone acetate for peak identification European Pharmacopoeia (EP) Reference Standard

Hydrocortisone acetate for peak identification

Hydrocortisone acetate for peak identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Hydrocortisone 21-acetate, 11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-acetate, 17-α-Hydroxycorticosterone acetate, 17-Hydroxycorticosterone 21-acetate, 21-Acetoxy-4-pregnene-11β,17α-diol-3,20-dione, 4-Pregnene-11β,17α,21-triol-3,20-dione 21-acetate
Empirical Formula (Hill Notation):
C23H32O6
CAS Number:
Molecular Weight:
404.50

Products

Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1