Hyaluronic acid sodium salt from bovine vitreous humor
Synonym(s):
Poly(β-glucuronic acid-[1→3]-β-N-acetylglucosamine-[1→4]), alternating
Linear Formula:
(C14H21NaNO11)n
CAS Number:
Products
Product Category
Brand
Biological Source
Color
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing