Skip to Content
Merck
Heptanoic acid analytical standard

Heptanoic acid

Heptanoic acid analytical standard
Synonym(s):
Enanthic acid, Oenanthic acid, Heptanoic acid
Linear Formula:
CH3(CH2)5COOH
CAS Number:
Molecular Weight:
130.18
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing