Skip to Content
IN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner CELLSTAR
®
microplate
Products
Product Category
plates (10)
Brand
Sigma-Aldrich (1)
Manufacturer
Wells
Color
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GN781090
Greiner CELLSTAR
®
microplate,
384 well, TC treated, black μClear
®
Expand
View Pricing
GN655090
Greiner CELLSTAR
®
microplate,
96 well, TC treated, black μClear
®
Expand
View Pricing
GN781091
Greiner CELLSTAR
®
microplate,
384 well, TC treated, black μClear
®
Expand
View Pricing
GN781080
Greiner CELLSTAR
®
microplate,
384 well, TC treated, white
Expand
View Pricing
GN784080
Greiner CELLSTAR
®
microplate,
384 well, Small Volume
™
, HiBase, white
Expand
View Pricing
GN655185
Greiner CELLSTAR
®
microplate,
96 well, TC treated, flat bottom, suspension, chimney style, clear, sterile
Expand
View Pricing
GN781182
Greiner CELLSTAR
®
microplate,
384 well, TC treated, clear
Expand
View Pricing
GN781098
Greiner CELLSTAR
®
microplate,
384 well, TC treated, white μClear
®
Expand
View Pricing
GN650180
Greiner CELLSTAR
®
microplate,
96 well, TC treated, clear
Expand
View Pricing
GN655180
Greiner CELLSTAR
®
microplate,
96 well, TC treated, clear
Expand
View Pricing