
Glipizide Related Compound A

Synonym(s):
N-{2-[4-(Aminosulfonyl)phenyl]ethyl}-5-methyl-2-pyrazinecarboxamide, 4-[2-(5-Methylpyrazine-2-carboxamido)ethyl]benzenesulfonamide
Empirical Formula (Hill Notation):
C14H16N4O3S
CAS Number:
Molecular Weight:
320.37
Products
Shipping
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2