Skip to Content
Merck
Gefitinib for system suitability European Pharmacopoeia (EP) Reference Standard

Gefitinib for system suitability

Gefitinib for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Gefitinib
Empirical Formula (Hill Notation):
C22H24ClFN4O3
CAS Number:
Molecular Weight:
446.90

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1