![(−)-Gallocatechin 3-gallate phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/221/520/ea581383-d61e-41f9-9c2b-4cbe48131f75/640/ea581383-d61e-41f9-9c2b-4cbe48131f75.png)
(−)-Gallocatechin 3-gallate
![(−)-Gallocatechin 3-gallate phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/221/520/ea581383-d61e-41f9-9c2b-4cbe48131f75/640/ea581383-d61e-41f9-9c2b-4cbe48131f75.png)
Synonym(s):
(−)-Gallocatechin gallate, (2S,3R)-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-(3,4,5-trihydroxybenzoate)
Empirical Formula (Hill Notation):
C22H18O11
CAS Number:
Molecular Weight:
458.37
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing