Skip to Content
Merck
Gallic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Gallic acid

Gallic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
3,4,5-Trihydroxybenzoic acid, Gallic acid
Linear Formula:
(HO)3C6H2CO2H
CAS Number:
Molecular Weight:
170.12
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Tissue
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4