Skip to Content
Merck
Fullerene-C60 98%

Fullerene-C60

Fullerene-C60 98%
Synonym(s):
Buckminsterfullerene
Empirical Formula (Hill Notation):
C60
CAS Number:
Molecular Weight:
720.64

Products

Shipping
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing