![Formaldehyde-2,4-dinitrophenylhydrazone BCR®, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/344/386/9e8c6b80-a251-4d29-b6b8-1da43c9303ac/640/9e8c6b80-a251-4d29-b6b8-1da43c9303ac.png)
Formaldehyde-2,4-dinitrophenylhydrazone
![Formaldehyde-2,4-dinitrophenylhydrazone BCR®, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/344/386/9e8c6b80-a251-4d29-b6b8-1da43c9303ac/640/9e8c6b80-a251-4d29-b6b8-1da43c9303ac.png)
Linear Formula:
H2C=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
210.15
Products
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing