Skip to Content
Merck
Fluoxetine Related Compound B United States Pharmacopeia (USP) Reference Standard

Fluoxetine Related Compound B

Fluoxetine Related Compound B United States Pharmacopeia (USP) Reference Standard
Synonym(s):
N-Methyl-N-(3-phenylpropyl)amine solution, N-Methyl-N-(3-phenylpropyl)amine
Empirical Formula (Hill Notation):
C10H15N
CAS Number:
Molecular Weight:
149.23

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade