Ethyl 4-hydroxybenzoate-ring-13C6 solution
Synonym(s):
4-Hydroxybenzoic acid ethyl ester-ring-13C6 solution, Ethyl paraben-ring-13C6 solution
Empirical Formula (Hill Notation):
13C6C3H10O3
Molecular Weight:
172.13
EC Number:
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing