Skip to Content
Merck
Erythrosin B analytical standard

Erythrosin B

Erythrosin B analytical standard
Synonym(s):
Erythrosin extra bluish, 2′,4′,5′,7′-Tetraiodofluorescein disodium salt, Acid Red 51, Iodoeosin
Empirical Formula (Hill Notation):
C20H6I4Na2O5
CAS Number:
Molecular Weight:
879.86
EC Number:

Products

Shipping
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2