
Endothelin 3 human, rat

Synonym(s):
ET-3
Empirical Formula (Hill Notation):
C121H168N26O33S4
CAS Number:
Molecular Weight:
2643.04
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-1 of 1