
D-Xylulose 5-phosphate lithium salt

Empirical Formula (Hill Notation):
C5H11O8P · xLi+
CAS Number:
Molecular Weight:
230.11 (free acid basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1