D(-)-N-Methylglucamine
Synonym(s):
N-Methyl-D-glucamine, 1-Deoxy-1-(methylamino)-D-glucitol, Meglumine
Empirical Formula (Hill Notation):
C7H17NO5
CAS Number:
Molecular Weight:
195.21
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing