Skip to Content
Merck
vibrant-m

(±)-Brompheniramine maleate salt

vibrant-m
Empirical Formula (Hill Notation):
C16H19BrN2 · C4H4O4
CAS Number:
Molecular Weight:
435.31

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1