Skip to Content
Merck
Diphenyl(trimethylsilyl)phosphine technical grade

Diphenyl(trimethylsilyl)phosphine

Diphenyl(trimethylsilyl)phosphine technical grade
Linear Formula:
(C6H5)2PSi(CH3)3
CAS Number:
Molecular Weight:
258.37

Products

Shipping
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing