Skip to Content
Merck
Diisobutylene technical, ≥90% (3 parts 2,4,4-trimethyl-1-pentene + 1 part 2,4,4-trimethyl-2-pentene, GC)

Diisobutylene

Diisobutylene technical, ≥90% (3 parts 2,4,4-trimethyl-1-pentene + 1 part 2,4,4-trimethyl-2-pentene, GC)
Synonym(s):
2,4,4-Trimethyl-1-pentene + 2,4,4-Trimethyl-2-pentene
Linear Formula:
(CH3)3CCH2C(CH3)=CH2+(CH3)3CCH=C(CH3)2
CAS Number:
Molecular Weight:
112.21

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Special Grade