Deflazacort
Synonym(s):
(11β,16β)-21-(acetyloxy)-11-hydroxy-2′-methyl-5′H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 11β,21-dihydroxy-2′-methyl-5′βH-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione 2′-acetate, DL-458-IT, L-5458, azacort, oxazacort, pregna-1,4-diene-11β,21-diol-3,20-dione[17α,16α-d]-2′-methyloxazoline 21-acetate
Empirical Formula (Hill Notation):
C25H31NO6
CAS Number:
Molecular Weight:
441.52
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing