Skip to Content
Merck
Cyclopamine hydrate ≥98% (HPLC)

Cyclopamine hydrate

Cyclopamine hydrate ≥98% (HPLC)
Synonym(s):
11-deoxojervine hydrate
Empirical Formula (Hill Notation):
C27H41NO2 · xH2O
CAS Number:
Molecular Weight:
411.62 (anhydrous basis)

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing