
Conjugated (10E,12Z)-Linoleic acid solution

Synonym(s):
(10E,12Z)-10,12-Octadecadienoic acid, 10E,Z12-CLA, Linoleic acid (10-trans, 12-cis)
Empirical Formula (Hill Notation):
C18H32O2
CAS Number:
Molecular Weight:
280.45
Products
Brand
Biological Source
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing