
Coniine hydrochloride

Synonym(s):
(±)-2-Propylpiperidine hydrochloride, DL-Coniin hydrochloride
Empirical Formula (Hill Notation):
C8H17N · HCl
CAS Number:
Molecular Weight:
163.69
Products
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing