
Clomifene citrate for ID and assay

Synonym(s):
Clomiphene citrate salt, 2-(4-[2-Chloro-1,2-diphenylethenyl]phenoxy)-N,N-diethylethanamine
Empirical Formula (Hill Notation):
C26H28ClNO · C6H8O7
CAS Number:
Molecular Weight:
598.08
Products
Formula Weight
Manufacturer
API Family
Showing 1-1 of 1
Compare
Product Number
Description
Pricing