Skip to Content
Merck
Cisplatin British Pharmacopoeia (BP) Reference Standard

Cisplatin

Cisplatin British Pharmacopoeia (BP) Reference Standard
Synonym(s):
cis-Diammineplatinum(II) dichloride, cis-Dichlorodiammine platinum(II), cis-Platinum(II) diammine dichloride, Cisplatin
Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05
EC Number:

Products

Shipping
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique