
Chlorphenamine N-Oxide Maleate

Empirical Formula (Hill Notation):
C20H23ClN2O5
CAS Number:
Molecular Weight:
406.86
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing