Skip to Content
Merck
Cetirizine Related Compound B certified reference material, pharmaceutical secondary standard

Cetirizine Related Compound B

Cetirizine Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
2-(4-Benzhydrylpiperazin-1-yl)ethan-1-ol dihydrochloride, 4-(Diphenylmethyl)-1-piperazineethanol dihydrochloride
Empirical Formula (Hill Notation):
C19H24N2O · 2HCl
CAS Number:
Molecular Weight:
369.33

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade