
Carboprost tromethamine

Synonym(s):
Carboprost trometamol, (5Z,9α,11α,13E,15S)-9,11,15-Trihydroxy-15- methylprosta-5,13-dien-1-oic acid with 2-amino-2-(hydroxymethyl)-1,3-propanediol (1:1), Carboprost tromethamine
Empirical Formula (Hill Notation):
C25H47NO8
CAS Number:
Molecular Weight:
489.64
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing