Calcium bromide
Synonym(s):
Hydrobromic acid calcium salt
Linear Formula:
CaBr2
CAS Number:
Molecular Weight:
199.89
Products
Shipping
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing