![Calcitonin Gene Related Peptide rat ≥95% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/images/652/096/6fc5b95a-e8c4-4112-b528-f6296d83fa14/640/6fc5b95a-e8c4-4112-b528-f6296d83fa14.jpg)
Calcitonin Gene Related Peptide rat
![Calcitonin Gene Related Peptide rat ≥95% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/images/652/096/6fc5b95a-e8c4-4112-b528-f6296d83fa14/640/6fc5b95a-e8c4-4112-b528-f6296d83fa14.jpg)
Synonym(s):
α-CGRP, CGRP-I
Empirical Formula (Hill Notation):
C162H262N50O52S2
CAS Number:
Molecular Weight:
3806.25
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing