Skip to Content
Merck
vibrant-m

Buserelin for NMR identification

vibrant-m
Synonym(s):
Buserelin acetate salt, des-Gly10-[D-Ser[t-Bu]6]-LH-RH ethylamide acetate salt, pGlu-His-Trp-Ser-Tyr-D-Ser(t-Bu)-Leu-Arg-Pro-NHEt acetate salt
Empirical Formula (Hill Notation):
C60H86N16O13 · C2H4O2
CAS Number:
Molecular Weight:
1299.48

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1