
Bromothymol blue

Synonym(s):
Bromophenol Blue solution, 3′,3′′,5′,5′′-Tetrabromophenolsulfonephthalein, Bromophenol Blue sodium salt
Empirical Formula (Hill Notation):
C19H9Br4NaO5S
CAS Number:
Molecular Weight:
691.94
Products
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing