Skip to Content
Merck
Bromophenol Blue sodium salt Dye content 90 %, ACS reagent

Bromophenol Blue sodium salt

Bromophenol Blue sodium salt Dye content 90 %, ACS reagent
Synonym(s):
3′,3′′,5′,5′′-Tetrabromophenolsulfophthalein sodium salt, OmniPur® Bromophenol Blue, Sodium Salt
Empirical Formula (Hill Notation):
C19H9Br4NaO5S
CAS Number:
Molecular Weight:
691.94
EC Number:

Products

Shipping
Product Category
Brand
Color
Formula Weight
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Special Grade
Technique