Skip to Content
IN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
SILBERBRAND volumetric flask
Products
Product Category
flasks (11)
Brand
Sigma-Aldrich (2)
Manufacturer
Capacity (mL)
Closure Type
Glassware Class
Package Size
Sterility
Style
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z326313
BRAND
®
SILBERBRAND volumetric flask,
volume 200 mL, accuracy: 0.25 mL
Expand
View Pricing
Z326321
BRAND
®
SILBERBRAND volumetric flask,
volume 250 mL, accuracy: 0.25 mL
Expand
View Pricing
Z326305
BRAND
®
SILBERBRAND volumetric flask,
volume 100 mL, accuracy: 0.15 mL
Expand
View Pricing
Z326372
BRAND
®
SILBERBRAND volumetric flask,
volume 5,000 mL, accuracy: 1.8 mL
Expand
View Pricing
Z326348
BRAND
®
SILBERBRAND volumetric flask,
volume 500 mL, accuracy: 0.4 mL
Expand
View Pricing
Z326364
BRAND
®
SILBERBRAND volumetric flask,
volume 2,000 mL, accuracy: 0.9 mL
Expand
View Pricing
Z326267
BRAND
®
SILBERBRAND volumetric flask,
volume 10 mL, accuracy: 0.06 mL
Expand
View Pricing
Z326356
BRAND
®
SILBERBRAND volumetric flask,
volume 1,000 mL, accuracy: 0.6 mL
Expand
View Pricing
Z326291
BRAND
®
SILBERBRAND volumetric flask,
volume 50 mL, accuracy: 0.09 mL
Expand
View Pricing
Z326283
BRAND
®
SILBERBRAND volumetric flask,
volume 25 mL, accuracy: 0.06 mL
Expand
View Pricing
Z326275
BRAND
®
SILBERBRAND volumetric flask,
volume 20 mL, accuracy: 0.06 mL
Expand
View Pricing