Skip to Content
Merck
Boc-D-Ala-OH ≥98.0% (TLC)

Boc-D-Ala-OH

Boc-D-Ala-OH ≥98.0% (TLC)
Synonym(s):
Boc-D-alanine, Boc-D-Ala-OH
Linear Formula:
CH3CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
189.21

Products

Shipping
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing