
Betamethasone benzoate

Synonym(s):
Betamethasone 17-benzoate
Empirical Formula (Hill Notation):
C29H33FO6
CAS Number:
Molecular Weight:
496.57
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing